(2-(Benzyloxy)-4-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

57166-500MG 57166-1G 57166-5G 57166-25G
77329-876EA 85.49 USD
77329-876 77342-222 77366-170 77378-060
(2-(Benzyloxy)-4-methylphenyl)methanol ≥97%
(2-(Benzyloxy)-4-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₂
MW: 228.286 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00661590
CAS Number: 117571-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR