2-Iodo-1-methoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

57158-500MG 57158-1G 57158-5G 57158-25G
77329-870EA 215.27 USD
77329-870 77353-834 77366-164 77387-944
2-Iodo-1-methoxynaphthalene ≥97%
2-Iodo-1-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉IO
Storage Temperature: Ambient
MDL Number: MFCD17012521
CAS Number: 151560-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR