Methyl 1-ethoxy-2-naphthoate ≥97%

Supplier: AOBChem USA

Warning

57148-500MG 57148-1G 57148-5G 57148-25G
77329-866EA 243.08 USD
77329-866 77353-830 77366-160 77387-940
Methyl 1-ethoxy-2-naphthoate ≥97%
Methyl 1-ethoxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₃
MW: 230.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758089
CAS Number: 2901100-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR