1-Ethoxy-2-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

57147-500MG 57147-1G 57147-5G 57147-25G
77329-864EA 239.99 USD
77329-864 77353-828 77366-158 77387-938
1-Ethoxy-2-naphthoic acid ≥97%
1-Ethoxy-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₃
MW: 216.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14646696
CAS Number: 54245-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR