(1-Ethoxynaphthalen-2-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

57145-500MG 57145-1G 57145-5G
77329-860EA 307.97 USD
77329-860 77353-824 77378-058
(1-Ethoxynaphthalen-2-yl)boronic acid ≥97%
(1-Ethoxynaphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃BO₃
MW: 216.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758087
CAS Number: 2243465-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR