(4-Iodo-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

57143-500MG 57143-1G 57143-5G 57143-25G
77329-856EA 145.23 USD
77329-856 77345-090 77366-152 77387-934
(4-Iodo-2-methylphenyl)(methyl)sulfane ≥97%
(4-Iodo-2-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₉IS
MW: 264.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564432
CAS Number: 2734779-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR