1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

57141-500MG 57141-1G 57141-5G 57141-25G
77329-852EA 185.4 USD
77329-852 77347-364 77366-150 77387-930
1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)ethanone ≥97%
1-(2-Fluoro-5-methyl-3-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD34758084
CAS Number: 2755716-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR