3-Acetyl-2-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57140-500MG 57140-1G 57140-5G 57140-25G
77329-850EA 187.46 USD
77329-850 77353-820 77366-148 77387-928
3-Acetyl-2-fluoro-5-methylbenzaldehyde ≥97%
3-Acetyl-2-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD34758083
CAS Number: 2383715-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR