Ethyl 3-methyl-4-(methylthio)benzoate ≥97%

Supplier: AOBChem USA

Warning

57124-500MG 57124-1G 57124-5G 57124-25G
77329-844EA 198.79 USD
77329-844 77353-818 77366-142 77387-924
Ethyl 3-methyl-4-(methylthio)benzoate ≥97%
Ethyl 3-methyl-4-(methylthio)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂S
MW: 210.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564431
CAS Number: 2734774-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR