2-Bromo-1-chloro-3-ethoxy-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

57111-500MG 57111-1G 57111-5G
77338-080EA 462.47 USD
77338-080 77353-814 77378-050
2-Bromo-1-chloro-3-ethoxy-4-fluorobenzene ≥97%
2-Bromo-1-chloro-3-ethoxy-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
Storage Temperature: Ambient
MDL Number: MFCD34758071
CAS Number: 2385361-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR