4-Bromo-2-methoxy-4'-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

57101-500MG 57101-1G 57101-5G 57101-25G
77329-842EA 145.23 USD
77329-842 77345-088 77366-140 77387-922
4-Bromo-2-methoxy-4'-methyl-1,1'-biphenyl ≥97%
4-Bromo-2-methoxy-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO
MW: 277.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564429
CAS Number: 2734779-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR