(4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

57100-500MG 57100-1G 57100-5G 57100-25G
77329-840EA 126.69 USD
77329-840 77344-148 77366-138 77378-040
(4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-yl)methanol ≥97%
(4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758063
CAS Number: 2755723-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR