4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

57099-500MG 57099-1G 57099-5G 57099-25G
77329-838EA 95.79 USD
77329-838 77342-566 77366-136 77378-038
4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Bromo-2'-methoxy-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
MW: 291.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564428
CAS Number: 2734773-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR