5-Chloro-2,4-difluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

57091-500MG 57091-1G 57091-5G 57091-25G
77329-836EA 225.57 USD
77329-836 77353-804 77366-134 77387-920
5-Chloro-2,4-difluoro-1,1'-biphenyl ≥95%
5-Chloro-2,4-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇ClF₂
MW: 224.634 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530578
CAS Number: 2734774-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR