2-Ethoxy-4-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

57087-500MG 57087-1G 57087-5G
77335-968EA 371.83 USD
77335-968 77353-802 77378-036
2-Ethoxy-4-fluoro-3-methylbenzoic acid ≥97%
2-Ethoxy-4-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758053
CAS Number: 2384050-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR