1-Bromo-4-fluoro-2-isopropoxy-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

57083-500MG 57083-1G 57083-5G 57083-25G
77329-832EA 207.03 USD
77329-832 77353-796 77366-130 77387-916
1-Bromo-4-fluoro-2-isopropoxy-3-methylbenzene ≥95%
1-Bromo-4-fluoro-2-isopropoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrFO
MW: 247.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758049
CAS Number: 2755716-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR