5-Ethyl-1-methyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

57073-500MG 57073-1G 57073-5G 57073-25G
77329-830EA 220.42 USD
77329-830 77353-794 77366-128 77387-914
5-Ethyl-1-methyl-1H-pyrazole-4-carboxylic acid ≥97%
5-Ethyl-1-methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08700652
CAS Number: 1007541-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR