5-Chloro-4-iodo-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

57052-500MG 57052-1G 57052-5G
77339-594EA 602.55 USD
77339-594 77353-792 77378-032
5-Chloro-4-iodo-2-methylbenzaldehyde ≥97%
5-Chloro-4-iodo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClIO
MW: 280.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758045
CAS Number: 2386284-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR