(4-Bromo-5-chloro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

57039-500MG 57039-1G 57039-5G
77337-444EA 414.06 USD
77337-444 77353-782 77378-022
(4-Bromo-5-chloro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(4-Bromo-5-chloro-2-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrClN₂O
MW: 331.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564425
CAS Number: 2734773-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR