6-Bromo-3-fluoro-2-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

57025-500MG 57025-1G 57025-25G 57025-5G
77329-828EA 296.64 USD
77329-828 77353-776 77387-912 77378-016
6-Bromo-3-fluoro-2-(methylthio)phenol ≥97%
6-Bromo-3-fluoro-2-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₇H₆BrFOS
Storage Temperature: Ambient
MDL Number: MFCD28739510
CAS Number: 1805419-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR