1-(2,4-Difluoro-3-iodophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

57011-500MG 57011-1G 57011-5G 57011-25G
77329-824EA 208.06 USD
77329-824 77353-772 77366-124 77387-908
1-(2,4-Difluoro-3-iodophenyl)ethanol ≥97%
1-(2,4-Difluoro-3-iodophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34530575
CAS Number: 2734774-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR