1-(3-Bromo-2,4-difluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

57010-500MG 57010-1G 57010-5G 57010-25G
77329-822EA 151.41 USD
77329-822 77345-428 77366-122 77387-906
1-(3-Bromo-2,4-difluorophenyl)ethanol ≥97%
1-(3-Bromo-2,4-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29472089
CAS Number: 1897340-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR