1-Chloro-3-fluoro-2-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56996-500MG 56996-1G 56996-5G
77329-820EA 326.51 USD
77329-820 77353-770 77378-014
1-Chloro-3-fluoro-2-iodo-5-methylbenzene ≥97%
1-Chloro-3-fluoro-2-iodo-5-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅ClFI
MW: 270.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28751667
CAS Number: 1805502-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR