(2-Chloro-6-fluoro-4-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56995-500MG 56995-1G 56995-5G 56995-25G
77329-818EA 145.23 USD
77329-818 77345-086 77366-120 77387-904
(2-Chloro-6-fluoro-4-methylphenyl)(methyl)sulfane ≥97%
(2-Chloro-6-fluoro-4-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752231
CAS Number: 1805227-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR