1-Chloro-2-iodo-5-methoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56993-500MG 56993-1G 56993-5G
77329-816EA 342.99 USD
77329-816 77353-768 77378-012
1-Chloro-2-iodo-5-methoxy-3-methylbenzene ≥97%
1-Chloro-2-iodo-5-methoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClIO
MW: 282.506 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758010
CAS Number: 2383652-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR