(2-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

56980-1G 56980-5G 56980-25G
77340-960EA 64.89 USD
77340-960 77366-118 77378-006
(2-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(2-Fluoro-5-methylphenyl)(4-methylpiperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₃H₁₇FN₂O
MW: 236.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17352672
CAS Number: 1275697-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR