(2-Fluoro-5-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
56976-500MG
56976-1G
56976-5G
56976-25G
77329-814EA
252.35
USD
77329-814
77353-762
77366-112
77387-902
(2-Fluoro-5-methylphenyl)(pyrrolidin-1-yl)methanone ≥97%
(2-Fluoro-5-methylphenyl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₄FNO MW: 207.244 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16783478 CAS Number: 1248696-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...