(2-Chloro-3-(trifluoromethyl)phenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

56954-1G 56954-5G 56954-25G
77341-038EA 66.95 USD
77341-038 77366-102 77377-992
(2-Chloro-3-(trifluoromethyl)phenyl)(morpholino)methanone ≥97%
(2-Chloro-3-(trifluoromethyl)phenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD34757996
CAS Number: 2755722-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR