2-Chloro-N,N-diethyl-3-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

56953-1G 56953-5G 56953-25G
77341-244EA 78.28 USD
77341-244 77366-100 77377-990
2-Chloro-N,N-diethyl-3-(trifluoromethyl)benzamide ≥97%
2-Chloro-N,N-diethyl-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClF₃NO
MW: 279.686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530568
CAS Number: 2734775-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR