(2-Chloro-3-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

56951-1G 56951-5G 56951-25G
77341-344EA 82.4 USD
77341-344 77366-098 77377-988
(2-Chloro-3-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone ≥95%
(2-Chloro-3-(trifluoromethyl)phenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD34564415
CAS Number: 2734774-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR