(4-Chloro-2,6-difluorophenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56942-500MG 56942-1G 56942-5G 56942-25G
77329-812EA 98.88 USD
77329-812 77342-720 77366-094 77377-984
(4-Chloro-2,6-difluorophenyl)(methyl)sulphane ≥97%
(4-Chloro-2,6-difluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₅ClF₂S
Storage Temperature: Ambient
MDL Number: MFCD28751310
CAS Number: 1805520-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR