1,3-Difluoro-2,5-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

56938-500MG 56938-1G 56938-5G 56938-25G
77329-810EA 145.23 USD
77329-810 77345-084 77366-092 77387-900
1,3-Difluoro-2,5-diiodobenzene ≥97%
1,3-Difluoro-2,5-diiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₂I₂
MW: 365.886 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30721696
CAS Number: 1239481-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR