(2,6-Difluoro-4-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56937-1G 56937-5G 56937-25G
77341-936EA 108.15 USD
77341-936 77366-090 77377-982
(2,6-Difluoro-4-iodophenyl)(methyl)sulfane ≥97%
(2,6-Difluoro-4-iodophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₅F₂IS
MW: 286.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757995
CAS Number: 2707649-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR