3'-Bromo-5'-isopropyl-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56907-500MG 56907-1G 56907-5G 56907-25G
77329-806EA 281.19 USD
77329-806 77353-756 77366-086 77387-896
3'-Bromo-5'-isopropyl-2-methyl-1,1'-biphenyl ≥97%
3'-Bromo-5'-isopropyl-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757982
CAS Number: 2755724-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR