3-Bromo-5-isopropyl-4'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

56906-500MG 56906-1G 56906-5G 56906-25G
77329-804EA 244.11 USD
77329-804 77353-754 77366-084 77387-894
3-Bromo-5-isopropyl-4'-methyl-1,1'-biphenyl ≥95%
3-Bromo-5-isopropyl-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇Br
MW: 289.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757981
CAS Number: 2755723-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR