3-Bromo-4'-chloro-5-isopropyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56904-500MG 56904-1G 56904-5G
77338-844EA 524.27 USD
77338-844 77353-752 77377-978
3-Bromo-4'-chloro-5-isopropyl-1,1'-biphenyl ≥97%
3-Bromo-4'-chloro-5-isopropyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₄BrCl
MW: 309.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757979
CAS Number: 2755719-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR