(5-Bromo-2-(methylthio)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

56897-500MG 56897-1G 56897-5G 56897-25G
77329-802EA 198.79 USD
77329-802 77353-746 77366-082 77387-892
(5-Bromo-2-(methylthio)phenyl)boronic acid ≥95%
(5-Bromo-2-(methylthio)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂S
Storage Temperature: Ambient
MDL Number: MFCD34757975
CAS Number: 2342602-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR