4-Chloro-7-fluoro-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

56894-500MG 56894-1G 56894-5G 56894-25G
77329-800EA 269.86 USD
77329-800 77353-744 77366-078 77387-890
4-Chloro-7-fluoro-1H-indazole ≥95%
4-Chloro-7-fluoro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
MW: 170.571 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749921
CAS Number: 1000341-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR