(3-Chloro-4-fluoro-5-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56890-500MG 56890-1G 56890-5G 56890-25G
77329-798EA 260.59 USD
77329-798 77353-740 77366-076 77387-888
(3-Chloro-4-fluoro-5-iodophenyl)(methyl)sulfane ≥97%
(3-Chloro-4-fluoro-5-iodophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564414
CAS Number: 2707648-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR