(3-Chloro-4-fluoro-5-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56889-500MG 56889-1G 56889-5G
77335-690EA 363.59 USD
77335-690 77353-738 77377-968
(3-Chloro-4-fluoro-5-methylphenyl)(methyl)sulfane ≥97%
(3-Chloro-4-fluoro-5-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752235
CAS Number: 1807268-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR