(3-Chloro-2-fluoro-5-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56888-500MG 56888-1G 56888-5G
77335-852EA 367.71 USD
77335-852 77353-736 77377-966
(3-Chloro-2-fluoro-5-(methylthio)phenyl)methanol ≥97%
(3-Chloro-2-fluoro-5-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈ClFOS
MW: 206.665 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757971
CAS Number: 2755722-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR