3-Chloro-2-fluoro-5-(methylthio)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

56884-500MG 56884-1G 56884-5G
77335-412EA 353.29 USD
77335-412 77353-734 77377-964
3-Chloro-2-fluoro-5-(methylthio)benzoic acid ≥95%
3-Chloro-2-fluoro-5-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂S
MW: 220.648 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22490189
CAS Number: 1824559-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR