4'-Bromo-2,3'-dimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

56879-500MG 56879-1G 56879-5G 56879-25G
77329-794EA 95.79 USD
77329-794 77342-564 77366-074 77377-958
4'-Bromo-2,3'-dimethyl-1,1'-biphenyl ≥95%
4'-Bromo-2,3'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Br
MW: 261.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530563
CAS Number: 2649019-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR