4'-Bromo-5-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥95%
Supplier: AOBChem USA
|
Warning
|
56878-500MG
56878-1G
56878-5G
56878-25G
77329-792EA
168.92
USD
77329-792
77346-282
77366-072
77387-884
4'-Bromo-5-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥95%
4'-Bromo-5-fluoro-2,3'-dimethyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrF Storage Temperature: Ambient |
MDL Number:
MFCD34530562 CAS Number: 2734776-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...