1-(5-Bromo-2,4-difluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

56877-500MG 56877-1G 56877-5G 56877-25G
77329-790EA 99.91 USD
77329-790 77342-834 77366-070 77377-956
1-(5-Bromo-2,4-difluorophenyl)ethanol ≥97%
1-(5-Bromo-2,4-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28131151
CAS Number: 1824270-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR