4'-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
56876-500MG
56876-1G
56876-5G
56876-25G
77329-788EA
164.8
USD
77329-788
77346-144
77366-068
77387-882
4'-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl ≥97%
4'-Bromo-4-fluoro-2,3'-dimethyl-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₂BrF MW: 279.147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530561 CAS Number: 2734776-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...