4'-Bromo-3'-methyl-2,4-bis(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56875-500MG 56875-1G 56875-5G 56875-25G
77329-786EA 187.46 USD
77329-786 77353-728 77366-066 77387-880
4'-Bromo-3'-methyl-2,4-bis(trifluoromethyl)-1,1'-biphenyl ≥97%
4'-Bromo-3'-methyl-2,4-bis(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₉BrF₆
MW: 383.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530560
CAS Number: 2734773-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR