4'-Bromo-2,3,3'-trimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

56872-500MG 56872-1G 56872-5G 56872-25G
77329-782EA 126.69 USD
77329-782 77344-144 77366-062 77377-952
4'-Bromo-2,3,3'-trimethyl-1,1'-biphenyl ≥95%
4'-Bromo-2,3,3'-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
Storage Temperature: Ambient
MDL Number: MFCD34530558
CAS Number: 2734773-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR