4'-Bromo-2,3',4-trimethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

56871-500MG 56871-1G 56871-5G 56871-25G
77329-780EA 130.81 USD
77329-780 77344-348 77366-060 77387-878
4'-Bromo-2,3',4-trimethyl-1,1'-biphenyl ≥95%
4'-Bromo-2,3',4-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530557
CAS Number: 2734775-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR