5-(4-Bromo-3-methylphenyl)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

56870-500MG 56870-1G 56870-5G 56870-25G
77329-778EA 187.46 USD
77329-778 77353-726 77366-058 77387-876
5-(4-Bromo-3-methylphenyl)-2-chloropyridine ≥97%
5-(4-Bromo-3-methylphenyl)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₂H₉BrClN
MW: 282.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28337838
CAS Number: 1359945-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR